Assay Details
| Gene Symbol: Wdr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181400.3 | NP_852065.2 | 15 | 3812 | 95 | |
| GenBank mRNA | AK028113.1 | - | 9 | 2203 | 95 | |
| BC040337.1 | - | 15 | 3673 | 3586908 | 95 | |
More Information
Set Membership: |
|

Wdr47
Genomic Map