Assay Details
| Gene Symbol: Coro1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011779.3 | NP_035909.2 | 11 | 3223 | 117 | |
| GenBank mRNA | AK077552.1 | - | 11 | 3238 | 117 | |
| AK082986.1 | - | 11 | 3225 | 117 | ||
| AK088101.1 | - | 11 | 3221 | 117 | ||
| AK151017.1 | - | 11 | 3212 | 117 | ||
| AK151515.1 | - | 2 | 1993 | 117 | ||
| AK151749.1 | - | 11 | 3212 | 117 | ||
| AK152246.1 | - | 2 | 1993 | 117 | ||
| AK159506.1 | - | 11 | 3215 | 117 | ||
| AK167089.1 | - | 11 | 3211 | 117 | ||
| AK169262.1 | - | 11 | 3204 | 117 | ||
| AK169404.1 | - | 11 | 3227 | 117 | ||
| BC009126.1 | - | 9 | 2918 | 117 | ||
| BC019444.1 | - | 11 | 3204 | 4167810 | 117 | |
| BC087553.1 | - | 11 | 3211 | 30460345 | 117 | |
| BC099671.1 | - | 11 | 3169 | 4918161 | 117 | |
More Information
Set Membership: |
|

Coro1c
Genomic Map