Assay Details
| Gene Symbol: Rpl13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009438.5 | NP_033464.2 | 8 | 715 | 71 | |
| GenBank mRNA | AK007609.1 | - | 8 | 705 | 71 | |
| AK077512.1 | - | 8 | 707 | 71 | ||
| AK146318.1 | - | 8 | 707 | 71 | ||
| AK170030.1 | - | 8 | 618 | 71 | ||
| BC034450.1 | - | 2 | 248 | 71 | ||
| BC042392.1 | - | 2 | 248 | 71 | ||
More Information
Set Membership: |
|

Rpl13a
Genomic Map