Assay Details
| Gene Symbol: Hic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178922.3 | NP_849253.2 | 3 | 2117 | 61 | |
| GenBank mRNA | AF117382.1 | - | 1 | 841 | 61 | |
| AJ868292.1 | - | 2 | 2081 | 61 | ||
| AK028357.1 | - | 2 | 1927 | 61 | ||
| AK129266.1 | - | 3 | 2103 | 61 | ||
| AK145386.1 | - | 1 | 1224 | 61 | ||
| AK156332.1 | - | 3 | 2133 | 61 | ||
| AK157276.1 | - | 3 | 2133 | 61 | ||
| BC065124.1 | - | 2116 | 30537019 | 61 | ||
More Information
Set Membership: |
|

Hic2
Genomic Map