Assay Details
| Gene Symbol: Cblb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033238.1 | NP_001028410.1 | 18 | 5720 | 96 | |
| GenBank mRNA | AK147367.1 | - | 18 | 5705 | 96 | |
| AK164519.1 | - | 1 | 2713 | 96 | ||
| BC035536.1 | - | 10 | 4480 | 96 | ||
More Information
Set Membership: |
|

Cblb
Genomic Map