Assay Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252193.1 | NP_001239122.1 | 8 | 2104 | 95 | |
| NM_025275.4 | NP_079551.3 | 9 | 1923 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252193.1 | NP_001239122.1 | 8 | 2104 | 95 | |
| NM_025275.4 | NP_079551.3 | 9 | 1923 | 95 | ||
Set Membership: |
|