Assay Details
| Gene Symbol: Ost4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134692.2 | NP_001128164.1 | 1 | 309 | 102 | |
| NM_024460.4 | NP_077780.3 | 2 | 181 | 102 | ||
| GenBank mRNA | AA563474.1 | - | 2 | 181 | 102 | |
| AK002224.2 | - | 2 | 156 | 102 | ||
| AK002453.1 | - | 2 | 103 | 102 | ||
| AK003569.1 | - | 2 | 160 | 102 | ||
| AK005413.1 | - | 2 | 104 | 102 | ||
| AK009384.1 | - | 116 | 102 | |||
| AK010297.1 | - | 1 | 245 | 102 | ||
| AK019285.1 | - | 2 | 141 | 102 | ||
| BC002177.1 | - | 1 | 246 | 3486998 | 102 | |
| BC082316.1 | - | 1 | 220 | 6814413 | 102 | |
| BU756604.1 | - | 2 | 102 | |||
| BY089912.1 | - | 1 | 243 | 102 | ||
| CA466367.1 | - | 2 | 139 | 102 | ||
| DV041491.1 | - | 222 | 102 | |||
More Information
Set Membership: |
|

Ost4
Genomic Map