Assay Details
| Gene Symbol: Gm12830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033617.1 | 3 | 413 | 92 | ||
| GenBank mRNA | AK038383.1 | - | 3 | 414 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12830| Gene Symbol: Gm12830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033617.1 | 3 | 413 | 92 | ||
| GenBank mRNA | AK038383.1 | - | 3 | 414 | 92 | |
Set Membership: |
|