Assay Details
| Gene Symbol: Ndufb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026610.1 | NP_080886.1 | 2 | 308 | 67 | |
| GenBank mRNA | AK009807.1 | - | 2 | 308 | 67 | |
| AK018707.1 | - | 2 | 308 | 67 | ||
| BC056229.1 | - | 2 | 309 | 6475281 | 67 | |
| BC087907.1 | - | 2 | 306 | 67 | ||
| BC096421.1 | - | 2 | 307 | 67 | ||
More Information
Set Membership: |
|

Ndufb4
Genomic Map