Assay Details
| Gene Symbol: Ccr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190337.1 | NP_001177266.1 | 1 - 2 | 59 | 63 | |
| GenBank mRNA | BC105669.1 | - | 1 - 2 | 52 | 40046795 | 63 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105669.1, NM_001190337.1 transcripts.
Set Membership: |
|

Ccr6
Genomic Map