Assay Details
| Gene Symbol: Ndrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195006.1 | NP_001181935.1 | 13 - 14 | 984 | 61 | |
| GenBank mRNA | AK090374.1 | - | 13 - 14 | 966 | 61 | |
| AK144294.1 | - | 13 - 14 | 967 | 61 | ||
| AK168873.1 | - | 13 - 14 | 968 | 61 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK129305.1 | - | 1279 | Chr. - - [-] on Build | 61 | ||
More Information
Set Membership: |
|

Ndrg4
Genomic Map