Assay Details
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206367.1 | NP_001193296.1 | 12 - 13 | 1622 | 97 | |
| NM_001206368.1 | NP_001193297.1 | 11 - 12 | 1538 | 97 | ||
| NM_001206369.1 | NP_001193298.1 | 11 - 12 | 1583 | 97 | ||
| NM_146120.4 | NP_666232.2 | 11 - 12 | 1645 | 97 | ||
| GenBank mRNA | AK076156.1 | - | 11 - 12 | 1585 | 97 | |
| AK089934.1 | - | 12 - 13 | 1577 | 97 | ||
| AK135632.1 | - | 11 - 12 | 1358 | 97 | ||
| AK143664.1 | - | 11 - 12 | 1612 | 97 | ||
| AK146969.1 | - | 12 - 13 | 1539 | 97 | ||
| AK148957.1 | - | 12 - 13 | 1602 | 97 | ||
| AK150359.1 | - | 12 - 13 | 1545 | 97 | ||
| AK151684.1 | - | 12 - 13 | 1564 | 97 | ||
| AK153029.1 | - | 12 - 13 | 1542 | 97 | ||
| AK166831.1 | - | 11 - 12 | 1558 | 97 | ||
| AK166932.1 | - | 12 - 13 | 1536 | 97 | ||
| AK167918.1 | - | 12 - 13 | 1552 | 97 | ||
| AK168461.1 | - | 12 - 13 | 1555 | 97 | ||
| AK168532.1 | - | 12 - 13 | 1556 | 97 | ||
| AK168707.1 | - | 11 - 12 | 1491 | 97 | ||
| AK168725.1 | - | 12 - 13 | 1539 | 97 | ||
| AK169041.1 | - | 12 - 13 | 1542 | 97 | ||
| AK169670.1 | - | 12 - 13 | 1567 | 97 | ||
| BC023143.1 | - | 11 - 12 | 1583 | 3711602 | 97 | |
| BC060377.1 | - | 12 - 13 | 1570 | 4983664 | 97 | |
| J04953.1 | - | 10 - 11 | 1432 | 97 | ||
More Information
Set Membership: |
|

Gsn
Genomic Map