Assay Details
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206367.1 | NP_001193296.1 | 7 - 8 | 959 | 75 | |
| NM_001206368.1 | NP_001193297.1 | 6 - 7 | 875 | 75 | ||
| NM_001206369.1 | NP_001193298.1 | 6 - 7 | 920 | 75 | ||
| NM_146120.4 | NP_666232.2 | 6 - 7 | 982 | 75 | ||
| GenBank mRNA | AK089934.1 | - | 7 - 8 | 914 | 75 | |
| AK135632.1 | - | 5 - 6 | 695 | 75 | ||
| AK143664.1 | - | 6 - 7 | 949 | 75 | ||
| AK146969.1 | - | 7 - 8 | 876 | 75 | ||
| AK148957.1 | - | 7 - 8 | 939 | 75 | ||
| AK150359.1 | - | 7 - 8 | 882 | 75 | ||
| AK151684.1 | - | 7 - 8 | 901 | 75 | ||
| AK153029.1 | - | 7 - 8 | 879 | 75 | ||
| AK166831.1 | - | 6 - 7 | 895 | 75 | ||
| AK166932.1 | - | 7 - 8 | 873 | 75 | ||
| AK167918.1 | - | 7 - 8 | 889 | 75 | ||
| AK168461.1 | - | 7 - 8 | 892 | 75 | ||
| AK168532.1 | - | 7 - 8 | 893 | 75 | ||
| AK168725.1 | - | 7 - 8 | 876 | 75 | ||
| AK168829.1 | - | 6 - 7 | 927 | 75 | ||
| AK169041.1 | - | 7 - 8 | 879 | 75 | ||
| AK169670.1 | - | 7 - 8 | 904 | 75 | ||
| AK178123.1 | - | 2 - 3 | 128 | 75 | ||
| BC023143.1 | - | 6 - 7 | 920 | 3711602 | 75 | |
| BC060377.1 | - | 7 - 8 | 907 | 4983664 | 75 | |
| J04953.1 | - | 5 - 6 | 769 | 75 | ||
More Information
Set Membership: |
|

Gsn
Genomic Map