Assay Details
| Gene Symbol: Fsbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256142.1 | NP_001243071.1 | 1 - 2 | 451 | 59 | |
| GenBank mRNA | AK053419.1 | - | 1 - 2 | 450 | 59 | |
| AK147819.1 | - | 1 - 2 | 452 | 59 | ||
More Information
Set Membership: |
|

Fsbp
Genomic Map