Assay Details
| Gene Symbol: Lef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276402.1 | NP_001263331.1 | 6 - 7 | 1934 | 64 | |
| NM_001276403.1 | NP_001263332.1 | 7 - 8 | 666 | 64 | ||
| NM_010703.4 | NP_034833.2 | 7 - 8 | 2018 | 64 | ||
| GenBank mRNA | AK028246.1 | - | 6 - 7 | 1556 | 64 | |
| AK034819.1 | - | 6 - 7 | 1407 | 64 | ||
| AK083328.1 | - | 7 - 8 | 661 | 64 | ||
| AK131979.1 | - | 6 - 7 | 1916 | 64 | ||
| AK158769.1 | - | 7 - 8 | 2000 | 64 | ||
| BC038305.1 | - | 7 - 8 | 801 | 64 | ||
| BC057543.1 | - | 6 - 7 | 1940 | 6401514 | 64 | |
| X58636.1 | - | 7 - 8 | 1832 | 64 | ||
More Information
Set Membership: |
|

Lef1
Genomic Map