Assay Details
| Gene Symbol: Ube2t | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278115.1 | NP_001265044.1 | 2 - 3 | 216 | 92 | |
| NM_026024.3 | NP_080300.1 | 2 - 3 | 393 | 92 | ||
| GenBank mRNA | AK012565.1 | - | 2 - 3 | 352 | 92 | |
| AK021213.1 | - | 2 - 3 | 382 | 92 | ||
| AK166098.1 | - | 2 - 3 | 363 | 92 | ||
| BC029213.1 | - | 2 - 3 | 342 | 3591557 | 92 | |
| CK334733.1 | - | 2 - 3 | 207 | 92 | ||
More Information
Set Membership: |
|

Ube2t
Genomic Map