Assay Details
| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285812.1 | NP_001272741.1 | 2 - 3 | 320 | 61 | |
| NM_001285814.1 | NP_001272743.1 | 3 - 4 | 385 | 61 | ||
| NM_001285815.1 | NP_001272744.1 | 3 - 4 | 403 | 61 | ||
| NM_026837.3 | NP_081113.1 | 2 - 3 | 299 | 61 | ||
| NM_029589.1 | NP_083865.1 | 3 - 4 | 221 | 61 | ||
| GenBank mRNA | AK004164.1 | - | 2 - 3 | 262 | 61 | |
| AK005104.1 | - | 2 - 3 | 206 | 61 | ||
| AK005885.1 | - | 3 - 4 | 225 | 61 | ||
| BC019937.1 | - | 2 - 3 | 225 | 4023311 | 61 | |
| BC020121.1 | - | 2 - 3 | 229 | 3991677 | 61 | |
| BC039805.1 | - | 1 - 2 | 604 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001285815.1 transcripts.
Set Membership: |
|

Tmem53
Genomic Map