Assay Details
| Gene Symbol: Adh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288578.1 | NP_001275507.1 | 6 - 7 | 894 | 71 | |
| NM_007410.3 | NP_031436.2 | 6 - 7 | 912 | 71 | ||
| GenBank mRNA | AK002790.1 | - | 6 - 7 | 834 | 71 | |
| AK076507.1 | - | 6 - 7 | 841 | 71 | ||
| AK146949.1 | - | 6 - 7 | 856 | 71 | ||
| AK159803.1 | - | 6 - 7 | 892 | 71 | ||
| BC062879.1 | - | 6 - 7 | 838 | 5691327 | 71 | |
| BC090978.1 | - | 6 - 7 | 835 | 30313517 | 71 | |
| M84147.1 | - | 6 - 7 | 862 | 71 | ||
More Information
Set Membership: |
|

Adh5
Genomic Map