Assay Details
| Gene Symbol: Gsta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077353.2 | NP_001070821.1 | 5 - 6 | 460 | 76 | |
| NM_001288617.1 | NP_001275546.1 | 5 - 6 | 505 | 76 | ||
| NM_010356.4 | NP_034486.2 | 4 - 5 | 365 | 76 | ||
| GenBank mRNA | AI047326.1 | - | 5 - 6 | 438 | 76 | |
| AI255699.1 | - | 4 - 5 | 361 | 76 | ||
| AK002478.1 | - | 3 - 4 | 313 | 76 | ||
| AK014076.1 | - | 4 - 5 | 338 | 76 | ||
| AK149543.1 | - | 4 - 5 | 330 | 76 | ||
| BC147272.1 | - | 4 - 5 | 328 | 9056327 | 76 | |
| BC147273.1 | - | 4 - 5 | 328 | 9056339 | 76 | |
| GQ227791.1 | - | 3 - 4 | 290 | 76 | ||
| M73483.1 | - | 3 - 4 | 296 | 76 | ||
| X65021.1 | - | 3 - 4 | 281 | 76 | ||
More Information
Set Membership: |
|

Gsta3
Genomic Map