Assay Details
| Gene Symbol: Tcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289468.1 | NP_001276397.1 | 2 - 3 | 381 | 61 | |
| GenBank mRNA | AK149361.1 | - | 2 - 3 | 363 | 61 | |
More Information
Set Membership: |
|

Tcl1
Genomic Map