Assay Details
| Gene Symbol: Med27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029615.1 | NP_083891.1 | 4 - 5 | 601 | 79 | |
| GenBank mRNA | AK009764.1 | - | 4 - 5 | 582 | 79 | |
| AK147181.1 | - | 4 - 5 | 598 | 79 | ||
More Information
Set Membership: |
|

Med27
Genomic Map