Assay Details
| Gene Symbol: Mrps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291270.1 | NP_001278199.1 | 3 - 4 | 329 | 84 | |
| NM_001291271.1 | NP_001278200.1 | 3 - 4 | 573 | 84 | ||
| NM_001291272.1 | NP_001278201.1 | 3 - 4 | 440 | 84 | ||
| NM_001291273.1 | NP_001278202.1 | 2 - 3 | 174 | 84 | ||
| NM_024174.6 | NP_077136.2 | 3 - 4 | 345 | 84 | ||
| NR_111902.1 | 2 - 3 | 269 | 84 | |||
| GenBank mRNA | AK009880.1 | - | 3 - 4 | 400 | 84 | |
| AK014375.2 | - | 3 - 4 | 315 | 84 | ||
| AK168051.1 | - | 3 - 4 | 539 | 84 | ||
| BC002155.1 | - | 3 - 4 | 392 | 84 | ||
| BC019980.1 | - | 3 - 4 | 315 | 4458402 | 84 | |
| BY238199.1 | - | 3 - 4 | 351 | 84 | ||
More Information
Set Membership: |
|

Mrps23
Genomic Map