Assay Details
| Gene Symbol: Usp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301414.1 | NP_001288343.1 | 5 - 6 | 1062 | 72 | |
| NM_146144.4 | NP_666256.2 | 5 - 6 | 1043 | 72 | ||
| GenBank mRNA | AK080882.1 | - | 5 - 6 | 1044 | 72 | |
| BC018179.1 | - | 5 - 6 | 923 | 3585191 | 72 | |
| BC020007.1 | - | 5 - 6 | 895 | 3980469 | 72 | |
| BC023812.1 | - | 2 - 3 | 217 | 72 | ||
More Information
Set Membership: |
|

Usp1
Genomic Map