Assay Details
| Gene Symbol: Gemin8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146238.3 | NP_666350.2 | 1 - 2 | 75 | 196 | |
| GenBank mRNA | AK037580.1 | - | 1 - 2 | 64 | 103 | |
| AK160583.1 | - | 1 - 2 | 76 | 196 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160583.1, NM_146238.3 transcripts.
Set Membership: |
|

Gemin8
Genomic Map