Assay Details
| Gene Symbol: Ffar3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033316.2 | NP_001028488.1 | 1 - 2 | 51 | 75 | |
| GenBank mRNA | AK148616.1 | - | 1 - 2 | 52 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148616.1, NM_001033316.2 transcripts.
Set Membership: |
|

Ffar3
Genomic Map