Assay Details
| Gene Symbol: Eva1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199210.1 | NP_001186139.1 | 6 - 7 | 1272 | 97 | |
| NM_027627.2 | NP_081903.1 | 5 - 6 | 1128 | 97 | ||
| GenBank mRNA | AF358257.1 | - | 6 - 7 | 999 | 97 | |
| AK016443.1 | - | 8 - 9 | 1268 | 97 | ||
| AK037486.1 | - | 4 - 5 | 983 | 97 | ||
| BC100350.1 | - | 5 - 6 | 874 | 30522196 | 97 | |
| BC132305.1 | - | 6 - 7 | 1272 | 40130582 | 97 | |
| BC144751.1 | - | 5 - 6 | 1074 | 97 | ||
| BC157969.1 | - | 4 - 5 | 822 | 9007291 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037486.1 transcripts.
Set Membership: |
|

Eva1c
Genomic Map