Assay Details
| Gene Symbol: Stk38l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172734.3 | NP_766322.1 | 10 - 11 | 1085 | 103 | |
| GenBank mRNA | AK045971.1 | - | 8 - 9 | 822 | 103 | |
| AK052288.1 | - | 10 - 11 | 1021 | 103 | ||
| AY223819.1 | - | 10 - 11 | 1007 | 103 | ||
| BC038621.1 | - | 3 - 4 | 235 | 103 | ||
| BC062170.1 | - | 9 - 10 | 979 | 30135051 | 103 | |
More Information
Set Membership: |
|

Stk38l
Genomic Map