Assay Details
| Gene Symbol: 1700084C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC172001.1 | - | 1 - 2 | 237 | 9054287 | 80 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700084C01Rik| Gene Symbol: 1700084C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC172001.1 | - | 1 - 2 | 237 | 9054287 | 80 |
Set Membership: |
|