Assay Details
| Gene Symbol: Cdkn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007670.4 | NP_031696.1 | 1 - 2 | 369 | 113 | |
| GenBank mRNA | AF059567.1 | - | 1 - 2 | 278 | 113 | |
| AK144132.1 | - | 1 - 2 | 379 | 113 | ||
| AK146668.1 | - | 1 - 2 | 330 | 113 | ||
| BC002010.1 | - | 1 - 2 | 283 | 3495097 | 113 | |
| DT919921.1 | - | 1 - 2 | 108 | 113 | ||
More Information
Set Membership: |
|

Cdkn2b
Genomic Map