Assay Details
| Gene Symbol: Defb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007843.3 | NP_031869.1 | 1 - 2 | 154 | 107 | |
| GenBank mRNA | AI956736.1 | - | 1 - 2 | 220 | 107 | |
| AK142544.1 | - | 1 - 2 | 135 | 107 | ||
| BC024380.1 | - | 1 - 2 | 110 | 4975536 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142544.1, BC024380.1, NM_007843.3 transcripts.
Set Membership: |
|

Defb1
Genomic Map