Assay Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008944.2 | NP_032970.2 | 3 - 4 | 283 | 90 | |
| GenBank mRNA | AF099733.1 | - | 3 - 4 | 250 | 90 | |
| AK012119.1 | - | 3 - 4 | 265 | 90 | ||
| AK012990.1 | - | 3 - 4 | 272 | 90 | ||
| AK035578.1 | - | 272 | 90 | |||
| AK136115.1 | - | 3 - 4 | 285 | 90 | ||
| AK151883.1 | - | 3 - 4 | 274 | 90 | ||
| AK152862.1 | - | 3 - 4 | 274 | 90 | ||
| AK154011.1 | - | 3 - 4 | 267 | 90 | ||
| AK167752.1 | - | 3 - 4 | 274 | 90 | ||
| BC026768.1 | - | 3 - 4 | 256 | 3156907 | 90 | |
| CJ052982.1 | - | 3 - 4 | 285 | 90 | ||
| X70303.1 | - | 3 - 4 | 327 | 90 | ||
More Information
Set Membership: |
|

Psma2
Genomic Map