Assay Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008944.2 | NP_032970.2 | 6 - 7 | 559 | 101 | |
| GenBank mRNA | AF099733.1 | - | 6 - 7 | 526 | 101 | |
| AK012119.1 | - | 6 - 7 | 541 | 101 | ||
| AK012990.1 | - | 6 - 7 | 548 | 101 | ||
| AK035578.1 | - | 548 | 101 | |||
| AK136115.1 | - | 6 - 7 | 561 | 101 | ||
| AK151883.1 | - | 6 - 7 | 550 | 101 | ||
| AK152862.1 | - | 6 - 7 | 550 | 101 | ||
| AK154011.1 | - | 6 - 7 | 543 | 101 | ||
| AK167752.1 | - | 6 - 7 | 550 | 101 | ||
| BC026768.1 | - | 6 - 7 | 532 | 3156907 | 101 | |
| BU755659.1 | - | 2 - 3 | 406 | 101 | ||
More Information
Set Membership: |
|

Psma2
Genomic Map