Assay Details
| Gene Symbol: Polr1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009087.2 | NP_033113.1 | 1 - 2 | 342 | 73 | |
| GenBank mRNA | AK002811.1 | - | 1 - 2 | 110 | 73 | |
| D86609.1 | - | 1 - 2 | 147 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002811.1, D86609.1, NM_009087.2 transcripts.
Set Membership: |
|

Polr1d
Genomic Map