Assay Details
| Gene Symbol: Tpd52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025261.1 | NP_001020432.1 | 3 - 4 | 468 | 86 | |
| NM_001025262.1 | NP_001020433.1 | 3 - 4 | 468 | 86 | ||
| NM_001025263.1 | NP_001020434.1 | 3 - 4 | 371 | 86 | ||
| NM_001025264.1 | NP_001020435.1 | 3 - 4 | 371 | 86 | ||
| NM_009412.2 | NP_033438.1 | 3 - 4 | 371 | 86 | ||
| GenBank mRNA | AK005456.1 | - | 3 - 4 | 352 | 86 | |
| AK032111.1 | - | 3 - 4 | 334 | 86 | ||
| AK150935.1 | - | 3 - 4 | 334 | 86 | ||
| AK151471.1 | - | 3 - 4 | 332 | 86 | ||
| AK151744.1 | - | 3 - 4 | 332 | 86 | ||
| AK152543.1 | - | 3 - 4 | 332 | 86 | ||
| AK153266.1 | - | 3 - 4 | 332 | 86 | ||
| AK153292.1 | - | 3 - 4 | 342 | 86 | ||
| AK161753.1 | - | 3 - 4 | 333 | 86 | ||
| AK168379.1 | - | 3 - 4 | 327 | 86 | ||
| AK168817.1 | - | 3 - 4 | 335 | 86 | ||
| AK169751.1 | - | 3 - 4 | 364 | 86 | ||
| AK170725.1 | - | 3 - 4 | 317 | 86 | ||
| AK171050.1 | - | 3 - 4 | 312 | 86 | ||
| AY048852.1 | - | 3 - 4 | 460 | 86 | ||
| BC002036.1 | - | 3 - 4 | 351 | 3593963 | 86 | |
| BC004068.1 | - | 3 - 4 | 3590969 | 86 | ||
| BC094018.1 | - | 3 - 4 | 327 | 3674995 | 86 | |
| BE634665.1 | - | 3 - 4 | 353 | 86 | ||
| BG404433.1 | - | 3 - 4 | 463 | 86 | ||
| CA480795.1 | - | 3 - 4 | 505 | 86 | ||
| U44426.1 | - | 3 - 4 | 300 | 86 | ||
More Information
Set Membership: |
|

Tpd52
Genomic Map