Assay Details
| Gene Symbol: Adora3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009631.4 | NP_033761.2 | 1 - 2 | 733 | 80 | |
| GenBank mRNA | AK013534.1 | - | 1 - 2 | 732 | 80 | |
| AK154312.1 | - | 1 - 2 | 735 | 80 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adenosine receptor signaling pathway
histamine secretion by mast cell
positive regulation of leukocyte migration
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of mast cell degranulation
positive regulation of inflammatory response
positive regulation of calcium-mediated signaling
positive regulation of mucus secretion
G-protein coupled adenosine receptor activity
adenosine receptor signaling pathway
histamine secretion by mast cell
positive regulation of leukocyte migration
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of mast cell degranulation
positive regulation of inflammatory response
positive regulation of calcium-mediated signaling
positive regulation of mucus secretion
G-protein coupled adenosine receptor activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972605 | AK154312.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972605 | BC100416.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972605 | NM_009631.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972605 | AK013534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972605 | NM_001174169.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135936 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972605 | AK154312.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972605 | BC100416.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972605 | NM_009631.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972605 | AK013534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972605 | NM_001174169.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P362981 | AK013534.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P31697 | L20331.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399860 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399853 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22399838 | - | - |
| Whole Mouse Genome G4122A | A_52_P31697 | L20331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P362981 | AK013534.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121269 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133063 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113267 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02386059 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03125155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03125143 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02871248 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03125150 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03125154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22399884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02457104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03125149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03192196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02558831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02923636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02923631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02923629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02519435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02923630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02311188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03125156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02558832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03125139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03103759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03125154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02923642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03125147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02395975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03125155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02395974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22399856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10495098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10495109 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430482_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429609_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495107 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10495097 | - | - |
| Mouse Genome 430 2.0 Array | 1430482_at | - | - |
| Mouse Genome 430 2.0 Array | 1430482_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11542_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11542_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300008853.mm.1 | NM_001174169.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008853.mm.1 | NM_009631.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008853.mm.1 | TC0300001096.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300008852.mm.1 | NM_009631.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008852.mm.1 | TC0300001096.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300008854.mm.1 | NM_001174169.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008854.mm.1 | NM_009631.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300008854.mm.1 | TC0300001096.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1430482_at | - | - |
| Mouse Expression Set 430 | 1430482_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430482_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430482_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429609_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429609_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 101190609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101190609 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 630333 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000630333 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226949 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2507802 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1226949 | - | - |
| mouse-6 v1.1 (Illumina) | scl22756.2.1_292-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0011542.2_271-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101190609 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 630333 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0011542.2_271-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630333 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101190609 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3061260 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226949 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2507802 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3061260 | - | - |

Adora3
Genomic Map