Assay Details
| Gene Symbol: Efna3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010108.1 | NP_034238.1 | 4 - 5 | 607 | 84 | |
| GenBank mRNA | BC107002.2 | - | 4 - 5 | 607 | 40045387 | 84 |
| BC125003.1 | - | 4 - 5 | 607 | 40045386 | 84 | |
More Information
Set Membership: |
|

Efna3
Genomic Map