Assay Details
| Gene Symbol: Gab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162477.1 | NP_001155949.1 | 9 - 10 | 1930 | 86 | |
| NM_010248.2 | NP_034378.2 | 9 - 10 | 1933 | 86 | ||
| GenBank mRNA | AB018414.1 | - | 9 - 10 | 1855 | 86 | |
| AF104244.1 | - | 9 - 10 | 1933 | 86 | ||
| BC103524.1 | - | 9 - 10 | 1905 | 40039291 | 86 | |
| BC103525.1 | - | 9 - 10 | 1908 | 40039280 | 86 | |
| BC103526.1 | - | 9 - 10 | 1908 | 40039281 | 86 | |
| BC103527.1 | - | 9 - 10 | 1908 | 40039282 | 86 | |
More Information
Set Membership: |
|

Gab2
Genomic Map