Assay Details
| Gene Symbol: Gbp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010259.2 | NP_034389.2 | 9 - 10 | 1539 | 66 | |
| GenBank mRNA | AK077796.1 | - | 9 - 10 | 1541 | 66 | |
| BC108990.2 | - | 1584 | 40047769 | 66 | ||
| EF494422.1 | - | 9 - 10 | 1539 | 66 | ||
| M55544.1 | - | 1523 | 66 | |||
| M63961.1 | - | 1617 | 66 | |||
More Information
Set Membership: |
|

Gbp2b
Genomic Map