Assay Details
| Gene Symbol: Hspa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010478.2 | NP_034608.2 | 1 | 2618 | 101 | |
| GenBank mRNA | AK159139.1 | - | 2586 | 101 | ||
| AK165042.1 | - | 1 | 2625 | 101 | ||
More Information
Set Membership: |
|

Hspa1b
Genomic Map