Assay Details
| Gene Symbol: Klf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010638.4 | NP_034768.2 | 1 - 2 | 1036 | 106 | |
| GenBank mRNA | AK028544.1 | - | 1 - 2 | 1039 | 106 | |
| BC106117.1 | - | 2 - 3 | 863 | 106 | ||
| BC138724.1 | - | 1 - 2 | 854 | 8861746 | 106 | |
| BC138725.1 | - | 1 - 2 | 849 | 8861747 | 106 | |
| CT010185.1 | - | 1 - 2 | 507 | 106 | ||
| Y14296.1 | - | 1 - 2 | 507 | 106 | ||
More Information
Set Membership: |
|

Klf9
Genomic Map