Assay Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011192.3 | NP_035322.1 | 8 - 9 | 732 | 108 | |
| GenBank mRNA | AK140597.1 | - | 8 - 9 | 753 | 108 | |
| AK150939.1 | - | 8 - 9 | 752 | 108 | ||
| AK156079.1 | - | 8 - 9 | 702 | 108 | ||
| AK171348.1 | - | 8 - 9 | 748 | 108 | ||
| BC015255.1 | - | 8 - 9 | 732 | 4221225 | 108 | |
| BC139818.1 | - | 8 - 9 | 680 | 40141268 | 108 | |
| CT010198.1 | - | 8 - 9 | 536 | 108 | ||
| D87911.1 | - | 8 - 9 | 655 | 108 | ||
More Information
Set Membership: |
|

Psme3
Genomic Map