Assay Details
| Gene Symbol: Sox14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011440.1 | NP_035570.1 | 1 | 423 | 112 | |
| GenBank mRNA | AK138208.1 | - | 1 | 315 | 112 | |
| BC055767.1 | - | 1 | 413 | 112 | ||
| BC069948.1 | - | 1 | 313 | 112 | ||
| BC083360.1 | - | 1 | 420 | 112 | ||
| BC100555.1 | - | 1 | 423 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138208.1, NM_011440.1 transcripts.
Set Membership: |
|

Sox14
Genomic Map