Assay Details
| Gene Symbol: Vax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011912.3 | NP_036042.1 | 2 - 3 | 466 | 60 | |
| GenBank mRNA | AB221556.1 | - | 2 - 3 | 437 | 60 | |
| AF028715.1 | - | 2 - 3 | 440 | 60 | ||
| BC090635.1 | - | 2 - 3 | 463 | 30942578 | 60 | |
| BC116389.1 | - | 2 - 3 | 458 | 40101824 | 60 | |
| BC116390.1 | - | 2 - 3 | 458 | 40101825 | 60 | |
| Y17792.1 | - | 2 - 3 | 466 | 60 | ||
More Information
Set Membership: |
|

Vax2
Genomic Map