Assay Details
| Gene Symbol: Tcl1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013773.2 | NP_038801.1 | 1 - 2 | 201 | 132 | |
| GenBank mRNA | AF195488.1 | - | 1 - 2 | 201 | 132 | |
| AK136181.1 | - | 1 - 2 | 217 | 132 | ||
| BC052337.1 | - | 1 - 2 | 237 | 30053367 | 132 | |
More Information
Set Membership: |
|

Tcl1b1
Genomic Map