Assay Details
| Gene Symbol: Magel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013779.2 | NP_038807.4 | 1 | 3910 | 108 | |
| GenBank mRNA | AJ243608.1 | - | 1 | 1425 | 108 | |
| AK082944.1 | - | 1 | 3883 | 108 | ||
| AK086725.1 | - | 1 | 3905 | 108 | ||
| BC054763.1 | - | 1 | 1202 | 108 | ||
| BC062913.1 | - | 3 | 3415 | 108 | ||
| BC071222.1 | - | 1 | 1688 | 108 | ||
More Information
Set Membership: |
|

Magel2
Genomic Map