Assay Details
| Gene Symbol: Peg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013788.2 | NP_038816.1 | 1 | 718 | 88 | |
| GenBank mRNA | AF148857.1 | - | 1 | 718 | 88 | |
| BC138242.1 | - | 1 | 686 | 8861261 | 88 | |
| BC138243.1 | - | 1 | 686 | 8861262 | 88 | |
More Information
Set Membership: |
|

Peg12
Genomic Map