Assay Details
| Gene Symbol: Rplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018853.3 | NP_061341.1 | 1 - 2 | 157 | 99 | |
| GenBank mRNA | AK007832.1 | - | 1 - 2 | 188 | 99 | |
| AK010656.1 | - | 1 - 2 | 189 | 99 | ||
| AK088070.1 | - | 1 - 2 | 187 | 99 | ||
| AK169347.1 | - | 1 - 2 | 187 | 99 | ||
| BC013252.1 | - | 1 - 2 | 122 | 99 | ||
| BC058685.1 | - | 1 - 2 | 199 | 6410847 | 99 | |
| BC091747.1 | - | 1 - 2 | 162 | 2655117 | 99 | |
| BC092088.1 | - | 1 - 2 | 161 | 5102711 | 99 | |
| BC092536.1 | - | 157 | 6774435 | 99 | ||
| U29402.1 | - | 1 - 2 | 185 | 99 | ||
More Information
Set Membership: |
|

Rplp1
Genomic Map