Assay Details
| Gene Symbol: Rplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018853.3 | NP_061341.1 | 2 - 3 | 230 | 88 | |
| GenBank mRNA | AK007832.1 | - | 2 - 3 | 261 | 88 | |
| AK010656.1 | - | 2 - 3 | 262 | 88 | ||
| AK088070.1 | - | 2 - 3 | 260 | 88 | ||
| AK169347.1 | - | 2 - 3 | 260 | 88 | ||
| BC013252.1 | - | 2 - 3 | 195 | 88 | ||
| BC058685.1 | - | 2 - 3 | 272 | 6410847 | 88 | |
| BC091747.1 | - | 2 - 3 | 235 | 2655117 | 88 | |
| BC092088.1 | - | 2 - 3 | 234 | 5102711 | 88 | |
| BC092536.1 | - | 230 | 6774435 | 88 | ||
| U29402.1 | - | 2 - 3 | 258 | 88 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567745 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567742 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567747 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567740 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567746 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740616 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140529 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155826 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P219603 | U29402.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26567747 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140529 | - | - |
| Whole Mouse Genome G4122A | A_51_P219603 | U29402.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114468 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07973952 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07759534 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07973961 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07973960 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567746 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567742 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567745 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567740 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26567747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26567686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07627972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07620628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08072652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08072644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07554504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08072640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07492787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07964798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07740944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07620635 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07674965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26567755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07492778 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581498 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581496 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10503834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581497 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10594250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10594249 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416277_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10503833 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10594248 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581495 | - | - |
| Mouse Genome 430 2.0 Array | 1416277_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1416277_a_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900020503.mm.1 | NM_018853.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900020503.mm.1 | NM_018853.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900020503.mm.1 | TC0900002450.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900020505.mm.1 | NM_018853.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900020505.mm.1 | NM_018853.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900020505.mm.1 | TC0900002450.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900020504.mm.1 | NM_018853.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900020504.mm.1 | NM_018853.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900020504.mm.1 | TC0900002450.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416277_PM_a_at | - | - |
| Mouse Expression Set 430 | 1416277_a_at | - | - |
| Murine Genome U74v2 Set | 100694_at | - | - |
| Murine Genome U74v2 Set | 161480_PM_i_at | - | - |
| Murine Genome U74v2 Set | 100694_PM_at | - | - |
| Murine Genome U74v2 Set | 161480_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416277_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416277_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416277_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416277_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104570068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104570068 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3140292 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003140292 | - | - |
| mouse-6 v1.1 (Illumina) | scl0003437.1_0-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_9256518-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104570068 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3140292 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003437.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104570068 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3140292 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2964873 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2964873 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2702561 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238874 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003437.1_0-S | - | - |

Rplp1
Genomic Map