Assay Details
| Gene Symbol: Psmg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019537.2 | NP_062410.1 | 3 - 4 | 407 | 101 | |
| GenBank mRNA | AJ238270.1 | - | 3 - 4 | 409 | 101 | |
| AK042203.1 | - | 3 - 4 | 418 | 101 | ||
| AK088118.1 | - | 3 - 4 | 420 | 101 | ||
| BC032965.1 | - | 3 - 4 | 400 | 1196077 | 101 | |
| BC060285.1 | - | 3 - 4 | 421 | 6508084 | 101 | |
More Information
Set Membership: |
|

Psmg1
Genomic Map