Assay Details
| Gene Symbol: Lpar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020028.3 | NP_064412.2 | 2 - 3 | 888 | 95 | |
| GenBank mRNA | AK051092.1 | - | 2 - 3 | 838 | 95 | |
| AK144861.1 | - | 2 - 3 | 889 | 95 | ||
| BC060131.1 | - | 2 - 3 | 1024 | 6409729 | 95 | |
| BC064676.1 | - | 2 - 3 | 1024 | 6409729 | 95 | |
More Information
Set Membership: |
|

Lpar2
Genomic Map